Tisk…

2,3-Difluoro-6-methoxybenzyl alcohol 97%

Dodavatel: Apollo Scientific

PC302660-10G PC302660-5G PC302660-1G
APOSPC302660-10GEA 9590 CZK
APOSPC302660-10G APOSPC302660-5G APOSPC302660-1G
2,3-Difluoro-6-methoxybenzyl alcohol 97%
2,3-Difluoro-6-methoxybenzyl alcohol
Vzorec: C₈H₈F₂O₂
MW: 174.145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04116021
CAS číslo: 773871-99-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR