Tisk…

2,3-Difluoro-6-methoxybenzoyl chloride 97%

Dodavatel: Apollo Scientific

PC302659-5G PC302659-1G
APOSPC302659-5GEA 5130 CZK
APOSPC302659-5G APOSPC302659-1G
2,3-Difluoro-6-methoxybenzoyl chloride 97%
2,3-Difluoro-6-methoxybenzoyl chloride
Vzorec: C₈H₅ClF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD04116020
CAS číslo: 886501-67-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR