2-Chloro-3-(trifluoromethyl)cinnamic acid 97%
Dodavatel: Apollo Scientific
PC302639-5G
PC302639-1G
APOSPC302639-5GEA
3580
CZK
APOSPC302639-5G
APOSPC302639-1G
2-Chloro-3-(trifluoromethyl)cinnamic acid 97%
2-Chloro-3-(trifluoromethyl)cinnamic acid
Vzorec:
C₁₀H₆ClF₃O₂ MW: 250,602 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03002851 CAS číslo: 682805-11-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...