Tisk…

5-Methyl-2-(trifluoromethyl)-DL-phenylalanine

Dodavatel: Apollo Scientific

PC302626-1G PC302626-5G PC302626-10G
APOSPC302626-1GEA 6390 CZK
APOSPC302626-1G APOSPC302626-5G APOSPC302626-10G
5-Methyl-2-(trifluoromethyl)-DL-phenylalanine
5-Methyl-2-(trifluoromethyl)-DL-phenylalanine
Vzorec: C₁₁H₁₂F₃NO₂
MW: 247,214 g/mol
Bod tání: 236 °C
Storage Temperature: Ambient
MDL Number: MFCD09832357
CAS číslo: 1256482-64-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR