5-Chloro-2-fluoro-DL-phenylglycine
Dodavatel: Apollo Scientific
PC302617-1G
PC302617-5G
APOSPC302617-1GEA
4530
CZK
APOSPC302617-1G
APOSPC302617-5G
5-Chloro-2-fluoro-DL-phenylglycine
5-Chloro-2-fluoro-DL-phenylglycine
Vzorec:
C₈H₇ClFNO₂ MW: 203,598 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04116048 CAS číslo: 1040198-59-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...