Tisk…

4-Fluoro-3,5-dimethylphenylacetic acid 97%

Dodavatel: Apollo Scientific
Varování

PC302595-1G PC302595-5G
APOSPC302595-1GEA 3230 CZK
APOSPC302595-1G APOSPC302595-5G
4-Fluoro-3,5-dimethylphenylacetic acid 97%
4-Fluoro-3,5-dimethylphenylacetic acid
Vzorec: C₁₀H₁₁FO₂
MW: 182,192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09832375
CAS číslo: 1000544-58-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR