Tisk…

4-Fluoro-2-methyl-DL-phenylglycine

Dodavatel: Apollo Scientific

PC302593-1G PC302593-5G
APOSPC302593-1GEA 5130 CZK
APOSPC302593-1G APOSPC302593-5G
4-Fluoro-2-methyl-DL-phenylglycine
4-Fluoro-2-methyl-DL-phenylglycine
Vzorec: C₉H₁₀FNO₂
MW: 183.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04116046
CAS číslo: 1039620-35-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR