Tisk…

4-Chloro-2-(trifluoromethyl)-DL-phenylalanine

Dodavatel: Apollo Scientific

PC302567-1G PC302567-5G PC302567-10G
APOSPC302567-1GEA 4200 CZK
APOSPC302567-1G APOSPC302567-5G APOSPC302567-10G
4-Chloro-2-(trifluoromethyl)-DL-phenylalanine
4-Chloro-2-(trifluoromethyl)-DL-phenylalanine
Vzorec: C₁₀H₉ClF₃NO₂
MW: 267.632 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115844
CAS číslo: 1259965-55-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR