Tisk…

3-Chloro-5-fluoro-DL-phenylglycine

Dodavatel: Apollo Scientific

PC302543-1G PC302543-5G
APOSPC302543-1GEA 4530 CZK
APOSPC302543-1G APOSPC302543-5G
3-Chloro-5-fluoro-DL-phenylglycine
3-Chloro-5-fluoro-DL-phenylglycine
Vzorec: C₈H₇ClFNO₂
MW: 203,598 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04116050
CAS číslo: 1038843-45-6
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR