Tisk…

3-Chloro-5-fluoro-4-methoxy-DL-phenylalanine

Dodavatel: Apollo Scientific

PC302539-1G PC302539-5G
APOSPC302539-1GEA 4860 CZK
APOSPC302539-1G APOSPC302539-5G
3-Chloro-5-fluoro-4-methoxy-DL-phenylalanine
3-Chloro-5-fluoro-4-methoxy-DL-phenylalanine
Vzorec: C₁₀H₁₁ClFNO₃
MW: 247,651 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09832235
CAS číslo: 1260005-16-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR