Tisk…

3-Chloro-5-(trifluoromethyl)benzamide 97%

Dodavatel: Apollo Scientific

PC302532-5G PC302532-1G
APOSPC302532-5GEA 4600 CZK
APOSPC302532-5G APOSPC302532-1G
3-Chloro-5-(trifluoromethyl)benzamide 97%
3-Chloro-5-(trifluoromethyl)benzamide
Vzorec: C₈H₅ClF₃NO
MW: 223,58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115853
CAS číslo: 886497-03-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR