Tisk…

3-Chloro-4-(trifluoromethoxy)phenylacetonitrile 97%

Dodavatel: Apollo Scientific

PC302523-1G PC302523-5G
APOSPC302523-1GEA 3310 CZK
APOSPC302523-1G APOSPC302523-5G
3-Chloro-4-(trifluoromethoxy)phenylacetonitrile 97%
3-Chloro-4-(trifluoromethoxy)phenylacetonitrile
Vzorec: C₉H₅ClF₃NO
Storage Temperature: Ambient
MDL Number: MFCD09832393
CAS číslo: 78240-45-6
UN: 3276
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR