3-Chloro-2-fluoro-DL-phenylalanine
Dodavatel: Apollo Scientific
PC302521-1G
PC302521-5G
PC302521-10G
APOSPC302521-1GEA
3310
CZK
APOSPC302521-1G
APOSPC302521-5G
APOSPC302521-10G
3-Chloro-2-fluoro-DL-phenylalanine
3-Chloro-2-fluoro-DL-phenylalanine
Vzorec:
C₉H₉ClFNO₂ MW: 217.625 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04116015 CAS číslo: 1259993-84-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...