Tisk…

3,6-Dichloro-2-fluorophenylacetonitrile 98%

Dodavatel: Apollo Scientific

PC302497-1G PC302497-5G PC302497-10G
APOSPC302497-1GEA 3870 CZK
APOSPC302497-1G APOSPC302497-5G APOSPC302497-10G
3,6-Dichloro-2-fluorophenylacetonitrile 98%
3,6-Dichloro-2-fluorophenylacetonitrile
Vzorec: C₈H₄Cl₂FN
MW: 204,028 g/mol
Bod tání: 74…76 °C
Storage Temperature: Ambient
MDL Number: MFCD09025390
CAS číslo: 916420-70-7
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR