Tisk…

3-(Trifluoromethoxy)-DL-phenylalanine

Dodavatel: Apollo Scientific

PC302493-1G PC302493-5G PC302493-250MG PC302493-25G
APOSPC302493-1GEA 8200 CZK
APOSPC302493-1G APOSPC302493-5G APOSPC302493-250MG APOSPC302493-25G
3-(Trifluoromethoxy)-DL-phenylalanine
3-(Trifluoromethoxy)-DL-phenylalanine
Vzorec: C₁₀H₁₀F₃NO₃
MW: 249.187 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04116035
CAS číslo: 439587-14-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR