Tisk…
3-(5-Fluoro-2-methoxyphenyl)propionic acid 98%
Dodavatel: Apollo Scientific
Synonyms: 3-(5-Fluoro-2-methoxyphenyl)propanoic acid
PC302490-1G PC302490-5G PC302490-10G PC302490-25G
APOSPC302490-1GEA 2780 CZK
APOSPC302490-1G APOSPC302490-5G APOSPC302490-10G APOSPC302490-25G
3-(5-Fluoro-2-methoxyphenyl)propionic acid 98%
3-(5-Fluoro-2-methoxyphenyl)propionic acid
Vzorec: C₁₀H₁₁FO₃
MW: 198,191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06823847
CAS číslo: 900021-53-6