3-(3,5-Difluoro-2-methoxyphenyl)propionic acid 97%
Dodavatel: Apollo Scientific
PC302478-1G
PC302478-5G
PC302478-10G
APOSPC302478-1GEA
3310
CZK
APOSPC302478-1G
APOSPC302478-5G
APOSPC302478-10G
3-(3,5-Difluoro-2-methoxyphenyl)propionic acid 97%
3-(3,5-Difluoro-2-methoxyphenyl)propionic acid
Vzorec:
C₁₀H₁₀F₂O₃ MW: 216,181 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09999481 CAS číslo: 1092460-64-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...