Tisk…

2-Ethoxy-3,5-difluorophenylacetonitrile 97%

Dodavatel: Apollo Scientific

PC302450-1G PC302450-5G PC302450-10G
APOSPC302450-1GEA 3870 CZK
APOSPC302450-1G APOSPC302450-5G APOSPC302450-10G
2-Ethoxy-3,5-difluorophenylacetonitrile 97%
2-Ethoxy-3,5-difluorophenylacetonitrile
Vzorec: C₁₀H₉F₂NO
MW: 197,181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258719
CAS číslo: 1017779-84-8
UN: 3276
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR