Tisk…

2-Chloro-6-fluoro-3-methoxy-DL-phenylalanine

Dodavatel: Apollo Scientific

PC302446-1G PC302446-5G PC302446-10G
APOSPC302446-1GEA 4530 CZK
APOSPC302446-1G APOSPC302446-5G APOSPC302446-10G
2-Chloro-6-fluoro-3-methoxy-DL-phenylalanine
2-Chloro-6-fluoro-3-methoxy-DL-phenylalanine
Vzorec: C₁₀H₁₁ClFNO₃
MW: 247,651 g/mol
Bod tání: 247…249 °C
Storage Temperature: Ambient
MDL Number: MFCD09832380
CAS číslo: 1256482-61-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR