Tisk…

2-Chloro-6-fluoro-3-methoxybenzamide 97%

Dodavatel: Apollo Scientific
Varování

PC302445-5G PC302445-1G
APOSPC302445-5GEA 4530 CZK
APOSPC302445-5G APOSPC302445-1G
2-Chloro-6-fluoro-3-methoxybenzamide 97%
2-Chloro-6-fluoro-3-methoxybenzamide
Vzorec: C₈H₇ClFNO₂
MW: 203,598 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660325
CAS číslo: 886761-58-6
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR