2,4,5-Trifluoro-DL-phenylglycine
Dodavatel: Apollo Scientific
PC302434-1G
PC302434-5G
APOSPC302434-1GEA
4960
CZK
APOSPC302434-1G
APOSPC302434-5G
2,4,5-Trifluoro-DL-phenylglycine
2,4,5-Trifluoro-DL-phenylglycine
Vzorec:
C₈H₆F₃NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD04116043 CAS číslo: 1043500-64-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...