Tisk…

2,3,6-Trifluoro-DL-phenylalanine

Dodavatel: Apollo Scientific

PC302426-1G PC302426-5G
APOSPC302426-1GEA 0 CZK
APOSPC302426-1G APOSPC302426-5G
2,3,6-Trifluoro-DL-phenylalanine
2,3,6-Trifluoro-DL-phenylalanine
Vzorec: C₉H₈F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD00454637
CAS číslo: 1214049-90-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR