Tisk…

2,3,4-Trifluoro-DL-phenylglycine

Dodavatel: Apollo Scientific

PC302424-1G PC302424-5G
APOSPC302424-1GEA 5960 CZK
APOSPC302424-1G APOSPC302424-5G
2,3,4-Trifluoro-DL-phenylglycine
2,3,4-Trifluoro-DL-phenylglycine
Vzorec: C₈H₆F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD04116042
CAS číslo: 1039627-62-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR