1-Chloro-2-(difluoromethoxy)-3-fluorobenzene
Dodavatel: Apollo Scientific
|
Varování
|
PC302418-1G
PC302418-5G
APOSPC302418-1GEA
3110
CZK
APOSPC302418-1G
APOSPC302418-5G
1-Chloro-2-(difluoromethoxy)-3-fluorobenzene
1-Chloro-2-(difluoromethoxy)-3-fluorobenzene
Vzorec:
C₇H₄ClF₃O Storage Temperature: Ambient |
MDL Number:
MFCD22580772 CAS číslo: 1136961-91-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...