6-Chloro-2-fluoro-3-methoxybenzyl alcohol 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC302417-5G
PC302417-1G
APOSPC302417-5GEA
3640
CZK
APOSPC302417-5G
APOSPC302417-1G
6-Chloro-2-fluoro-3-methoxybenzyl alcohol 98%
6-Chloro-2-fluoro-3-methoxybenzyl alcohol
Vzorec:
C₈H₈ClFO₂ MW: 190.599 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04115949 CAS číslo: 886499-68-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...