4-Chloro-3-(trifluoromethyl)benzamide 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC302410-10G
PC302410-5G
PC302410-1G
APOSPC302410-10GEA
1540
CZK
APOSPC302410-10G
APOSPC302410-5G
APOSPC302410-1G
4-Chloro-3-(trifluoromethyl)benzamide 97%
4-Chloro-3-(trifluoromethyl)benzamide
Vzorec:
C₈H₅ClF₃NO Storage Temperature: Ambient |
MDL Number:
MFCD04115832 CAS číslo: 62584-23-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...