Tisk…

4-Chloro-2-(trifluoromethyl)benzamide 97%

Dodavatel: Apollo Scientific

PC302407-5G PC302407-1G
APOSPC302407-5GEA 3940 CZK
APOSPC302407-5G APOSPC302407-1G
4-Chloro-2-(trifluoromethyl)benzamide 97%
4-Chloro-2-(trifluoromethyl)benzamide
Vzorec: C₈H₅ClF₃NO
MW: 223,58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115843
CAS číslo: 886496-79-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR