Tisk…

3-Chloro-5-fluorobenzoyl chloride

Dodavatel: Apollo Scientific

PC302400-10G PC302400-5G PC302400-1G
APOSPC302400-10GEA 10350 CZK
APOSPC302400-10G APOSPC302400-5G APOSPC302400-1G
3-Chloro-5-fluorobenzoyl chloride
3-Chloro-5-fluorobenzoyl chloride
Vzorec: C₇H₃Cl₂FO
MW: 193,003 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115835
CAS číslo: 886496-62-4
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR