Tisk…

5,6-Difluoroindole-2-carboxylic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 5,6-Difluoro-1H-indole-2-carboxylic acid

PC3024-1G PC3024-5G
APOSPC3024-1GEA 2530 CZK
APOSPC3024-1G APOSPC3024-5G
5,6-Difluoroindole-2-carboxylic acid 97%
5,6-Difluoroindole-2-carboxylic acid
Vzorec: C₉H₅F₂NO₂
MW: 197,138 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01863160
CAS číslo: 169674-35-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR