Tisk…
2-Chloro-6-fluoro-3-methoxybenzyl alcohol 97%
Dodavatel: Apollo Scientific
Synonyms: (2-Chloro-6-fluoro-3-methoxyphenyl)methanol
PC302386-5G PC302386-1G
APOSPC302386-5GEA 8700 CZK
APOSPC302386-5G APOSPC302386-1G
2-Chloro-6-fluoro-3-methoxybenzyl alcohol 97%
2-Chloro-6-fluoro-3-methoxybenzyl alcohol
Vzorec: C₈H₈ClFO₂
MW: 190,599 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115944
CAS číslo: 886499-49-6