Tisk…

2-Chloro-6-fluoro-3-methoxybenzoyl chloride 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC302385-5G PC302385-10G PC302385-1G
APOSPC302385-5GEA 5460 CZK
APOSPC302385-5G APOSPC302385-10G APOSPC302385-1G
2-Chloro-6-fluoro-3-methoxybenzoyl chloride 98%
2-Chloro-6-fluoro-3-methoxybenzoyl chloride
Vzorec: C₈H₅Cl₂FO₂
MW: 223,029 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115943
CAS číslo: 886499-44-1
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR