2-Chloro-6-(trifluoromethyl)benzyl alcohol
Dodavatel: Apollo Scientific
|
Varování
|
PC302384-10G
PC302384-5G
APOSPC302384-10GEA
0
CZK
APOSPC302384-10G
APOSPC302384-5G
2-Chloro-6-(trifluoromethyl)benzyl alcohol
2-Chloro-6-(trifluoromethyl)benzyl alcohol
Vzorec:
C₈H₆ClF₃O MW: 210,581 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00049315 CAS číslo: 886500-21-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...