Tisk…

5-Methyl-2-(trifluoromethyl)cinnamic acid 97%

Dodavatel: Apollo Scientific

PC302364-1G PC302364-5G
APOSPC302364-1GEA 2650 CZK
APOSPC302364-1G APOSPC302364-5G
5-Methyl-2-(trifluoromethyl)cinnamic acid 97%
5-Methyl-2-(trifluoromethyl)cinnamic acid
Vzorec: C₁₁H₉F₃O₂
MW: 230,183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09832354
CAS číslo: 1017779-24-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR