2-Fluoro-4-(pentafluorosulphur)benzenesulphonyl chloride 97%
Dodavatel: Apollo Scientific
PC302336-1G
PC302336-250MG
APOSPC302336-1GEA
54550
CZK
APOSPC302336-1G
APOSPC302336-250MG
2-Fluoro-4-(pentafluorosulphur)benzenesulphonyl chloride 97%
2-Fluoro-4-(pentafluorosulphur)benzenesulphonyl chloride
Vzorec:
C₆H₃ClF₆O₂S₂ Storage Temperature: Ambient |
MDL Number:
MFCD28054239 CAS číslo: 1706462-71-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...