4-Chloro-2-(trifluoromethyl)benzoyl chloride 98%
Dodavatel: Apollo Scientific
PC302321-25G
PC302321-5G
PC302321-1G
APOSPC302321-25GEA
14120
CZK
APOSPC302321-25G
APOSPC302321-5G
APOSPC302321-1G
4-Chloro-2-(trifluoromethyl)benzoyl chloride 98%
4-Chloro-2-(trifluoromethyl)benzoyl chloride
Vzorec:
C₈H₃Cl₂F₃O MW: 243,01 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04115838 CAS číslo: 98187-13-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...