3-Methyl-5-(trifluoromethyl)cinnamic acid 97%
Dodavatel: Apollo Scientific
PC302313-1G
PC302313-5G
APOSPC302313-1GEA
4530
CZK
APOSPC302313-1G
APOSPC302313-5G
3-Methyl-5-(trifluoromethyl)cinnamic acid 97%
3-Methyl-5-(trifluoromethyl)cinnamic acid
Vzorec:
C₁₁H₉F₃O₂ MW: 230,183 g/mol Bod tání: 170…173 °C Storage Temperature: Ambient |
MDL Number:
MFCD11519311 CAS číslo: 1092460-62-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...