2-Methoxy-5-(trifluoromethyl)benzylamine 97%
Dodavatel: Apollo Scientific
PC302257-1G
PC302257-5G
APOSPC302257-1GEA
4570
CZK
APOSPC302257-1G
APOSPC302257-5G
2-Methoxy-5-(trifluoromethyl)benzylamine 97%
2-Methoxy-5-(trifluoromethyl)benzylamine
Vzorec:
C₉H₁₀F₃NO MW: 205,177 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09832242 CAS číslo: 400771-42-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...