Tisk…

4-Fluoro-3-(trifluoromethoxy)cinnamic acid 98%

Dodavatel: Apollo Scientific

PC302213-1G PC302213-5G
APOSPC302213-1GEA 4200 CZK
APOSPC302213-1G APOSPC302213-5G
4-Fluoro-3-(trifluoromethoxy)cinnamic acid 98%
4-Fluoro-3-(trifluoromethoxy)cinnamic acid
Vzorec: C₁₀H₆F₄O₃
Storage Temperature: Ambient
MDL Number: MFCD06660259
CAS číslo: 886501-38-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR