Tisk…

4-Chloro-3-(trifluoromethoxy)phenylacetonitrile 97%

Dodavatel: Apollo Scientific

Synonyms: 2-(4-Chloro-3-(trifluoromethoxy)phenyl)acetonitrile

PC302208-1G PC302208-5G PC302208-10G PC302208-25G
APOSPC302208-1GEA 1080 CZK
APOSPC302208-1G APOSPC302208-5G APOSPC302208-10G APOSPC302208-25G
4-Chloro-3-(trifluoromethoxy)phenylacetonitrile 97%
4-Chloro-3-(trifluoromethoxy)phenylacetonitrile
Vzorec: C₉H₅ClF₃NO
MW: 235,59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660250
CAS číslo: 886500-97-6
UN: 3276
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR