Tisk…

3,5-Difluoro-2-methoxyphenylacetonitrile 98%

Dodavatel: Apollo Scientific

PC302184-1G PC302184-5G
APOSPC302184-1GEA 2980 CZK
APOSPC302184-1G APOSPC302184-5G
3,5-Difluoro-2-methoxyphenylacetonitrile 98%
3,5-Difluoro-2-methoxyphenylacetonitrile
Vzorec: C₉H₇F₂NO
MW: 183,155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660334
CAS číslo: 886761-64-4
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR