Tisk…

4-Chloro-2,6-difluoro-DL-phenylalanine

Dodavatel: Apollo Scientific

PC302145-1G PC302145-5G
APOSPC302145-1GEA 3150 CZK
APOSPC302145-1G APOSPC302145-5G
4-Chloro-2,6-difluoro-DL-phenylalanine
4-Chloro-2,6-difluoro-DL-phenylalanine
Vzorec: C₉H₈ClF₂NO₂
MW: 235,615 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660351
CAS číslo: 1259973-46-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR