Tisk…

3-Fluoro-5-methyl-DL-phenylalanine

Dodavatel: Apollo Scientific

PC302144-1G PC302144-5G
APOSPC302144-1GEA 3640 CZK
APOSPC302144-1G APOSPC302144-5G
3-Fluoro-5-methyl-DL-phenylalanine
3-Fluoro-5-methyl-DL-phenylalanine
Vzorec: C₁₀H₁₂FNO₂
MW: 197,206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05227967
CAS číslo: 603106-28-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR