3-Fluoro-5-(trifluoromethyl)-DL-phenylalanine
Dodavatel: Apollo Scientific
PC302143-1G
PC302143-5G
PC302143-10G
APOSPC302143-1GEA
3640
CZK
APOSPC302143-1G
APOSPC302143-5G
APOSPC302143-10G
3-Fluoro-5-(trifluoromethyl)-DL-phenylalanine
3-Fluoro-5-(trifluoromethyl)-DL-phenylalanine
Vzorec:
C₁₀H₉F₄NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD04115997 CAS číslo: 1259992-93-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...