Tisk…

3-Fluoro-2-methyl-DL-phenylalanine

Dodavatel: Apollo Scientific

PC302140-1G PC302140-5G
APOSPC302140-1GEA 3310 CZK
APOSPC302140-1G APOSPC302140-5G
3-Fluoro-2-methyl-DL-phenylalanine
3-Fluoro-2-methyl-DL-phenylalanine
Vzorec: C₁₀H₁₂FNO₂
MW: 197,206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660341
CAS číslo: 1260004-83-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR