Tisk…

3-Chloro-2,6-difluoro-DL-phenylalanine

Dodavatel: Apollo Scientific

PC302138-1G PC302138-5G PC302138-10G
APOSPC302138-1GEA 3310 CZK
APOSPC302138-1G APOSPC302138-5G APOSPC302138-10G
3-Chloro-2,6-difluoro-DL-phenylalanine
3-Chloro-2,6-difluoro-DL-phenylalanine
Vzorec: C₉H₈ClF₂NO₂
MW: 235.615 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04116006
CAS číslo: 1260006-14-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR