Tisk…

3-Chloro-2,4-difluoro-DL-phenylalanine

Dodavatel: Apollo Scientific

PC302137-1G PC302137-5G PC302137-10G
APOSPC302137-1GEA 4470 CZK
APOSPC302137-1G APOSPC302137-5G APOSPC302137-10G
3-Chloro-2,4-difluoro-DL-phenylalanine
3-Chloro-2,4-difluoro-DL-phenylalanine
Vzorec: C₉H₈ClF₂NO₂
MW: 235,615 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660336
CAS číslo: 1259964-91-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR