2-Chloro-4-fluoro-DL-phenylalanine
Dodavatel: Apollo Scientific
PC302127-1G
PC302127-5G
PC302127-10G
APOSPC302127-1GEA
3310
CZK
APOSPC302127-1G
APOSPC302127-5G
APOSPC302127-10G
2-Chloro-4-fluoro-DL-phenylalanine
2-Chloro-4-fluoro-DL-phenylalanine
Vzorec:
C₉H₉ClFNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD06660324 CAS číslo: 754152-25-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...