Tisk…

4-Chloro-2-fluorobenzamide 95%

Dodavatel: Apollo Scientific
Varování

PC302104-25G PC302104-5G PC302104-1G
APOSPC302104-25GEA 2570 CZK
APOSPC302104-25G APOSPC302104-5G APOSPC302104-1G
4-Chloro-2-fluorobenzamide 95%
4-Chloro-2-fluorobenzamide
Vzorec: C₇H₅ClFNO
MW: 173,572 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115956
CAS číslo: 104326-93-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR