2,4,6-Trifluoro-3-methoxybenzaldehyde 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Methoxy-2,4,6-trifluorobenzaldehyde
PC302070-1G
PC302070-5G
PC302070-10G
APOSPC302070-1GEA
2070
CZK
APOSPC302070-1G
APOSPC302070-5G
APOSPC302070-10G
2,4,6-Trifluoro-3-methoxybenzaldehyde 98%
2,4,6-Trifluoro-3-methoxybenzaldehyde
Vzorec:
C₈H₅F₃O₂ MW: 190,119 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06660224 CAS číslo: 886499-89-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...