Tisk…

3-Fluoro-5-methylcinnamic acid 97%

Dodavatel: Apollo Scientific

PC302056-1G PC302056-5G
APOSPC302056-1GEA 1660 CZK
APOSPC302056-1G APOSPC302056-5G
3-Fluoro-5-methylcinnamic acid 97%
3-Fluoro-5-methylcinnamic acid
Vzorec: C₁₀H₉FO₂
MW: 180,176 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06205749
CAS číslo: 773132-32-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR